Learn More
Sigma Aldrich Fine Chemicals Biosciences TrizmaR hydrochloride reag25KG
Catalog No. 502914289
Encompass
Trizma(R) hydrochloride is mainly required for preparation of buffer at physiological range of 7.3 to 7.5. The prepared buffer is compatible with biological fluids. It is of importance to laboratories as a standard pH SOLUTIONon.
Catalog No. 50-291-4289 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T325325KG
Trizma(R) hydrochloride is mainly required for preparation of buffer at physiological range of 7.3 to 7.5. The prepared buffer is compatible with biological fluids. It is of importance to laboratories as a standard pH SOLUTIONon.