Learn More
Sigma Aldrich Fine Chemicals Biosciences TrizmaR hydrochloride reag50KG
Catalog No. 502914337
Encompass
Trizma(R) hydrochloride is mainly required for preparation of buffer at physiological range of 7.3 to 7.5. The prepared buffer is compatible with biological fluids. It is of importance to laboratories as a standard pH SOLUTIONon.
Catalog No. 50-291-4337 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T325350KG
Add to Cart
Add to Cart
Request a Quote
Trizma(R) hydrochloride is mainly required for preparation of buffer at physiological range of 7.3 to 7.5. The prepared buffer is compatible with biological fluids. It is of importance to laboratories as a standard pH SOLUTIONon.