Learn More
Sigma Aldrich Fine Chemicals Biosciences TrizmaR hydrochloride solu
Catalog No. 502897080
Encompass
A series of Pre-mixed SOLUTIONons of TRIZMA Base and TRIZMA HCl to provide commonly used pH values for Tris buffers. No mixing or pH adjustment necessary. Guaranteed accuracy /- 0.1 pH units.
Catalog No. 50-289-7080 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T2194100ML
A series of Pre-mixed SOLUTIONons of TRIZMA Base and TRIZMA HCl to provide commonly used pH values for Tris buffers. No mixing or pH adjustment necessary. Guaranteed accuracy /- 0.1 pH units.