Learn More
Sigma Aldrich Fine Chemicals Biosciences TrizmaR hydrochloride solu
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T2819100ML
Encompass
Tris hydrochloride is used to prepare buffer mainly at a physiological range of 7.3 to 7.5. This buffer is well-suited with biological fluids. It is used in laboratories as primary pH standard in clinical laboratories.
Catalog No. 50-290-1182