Learn More
Sigma Aldrich Fine Chemicals Biosciences TrizmaR hydrochloride solu
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T1819100ML
Encompass
A series of Pre-mixed SOLUTIONons of TRIZMA Base and TRIZMA HCl to provide commonly used pH values for Tris buffers. No mixing or pH adjustment necessary. Guaranteed accuracy /- 0.1 pH units.
Catalog No. 50-290-1838