Learn More
Sigma Aldrich Fine Chemicals Biosciences TrizmaR hydrochloride solu1L
Supplier: Sigma Aldrich Fine Chemicals Biosciences T28191L
Tris hydrochloride is used to prepare buffer mainly at a physiological range of 7.3 to 7.5. This buffer is well-suited with biological fluids. It is used in laboratories as primary pH standard in clinical laboratories.