Learn More
Biotang Inc Trizyc 24-Well Plate 15ml
Catalog No. 502741235 Shop All BIOTANG Inc Products
Encompass_Preferred
Polypropylene 24-well deep w5mL. Adhesive sealing film, Hell rectangular plate. Square Well, Conical V shape bottom. Pre-sterilized. Volume: 1eat sealable film, Silicone sealing mat with plate.
Catalog No. 50-274-1235 Supplier Biotang Inc Supplier No. P15SQV24
Add to Cart
Add to Cart
Polypropylene 24-well deep w5mL. Adhesive sealing film, Hell rectangular plate. Square Well, Conical V shape bottom. Pre-sterilized. Volume: 1eat sealable film, Silicone sealing mat with plate.