Learn More
Biotang Inc Trizyc 24-Well Tip Comb
Supplier:  Biotang Inc PTB24
Encompass_Preferred
Kingfisher Flex-Compatible 24-Well Magnetic Tip Comb. High quality polypropylene (PP), good chemical resistance and heat-resistance. Standard size, suitable for common pipettes on the market. Optimize the pore size to ensure smooth sample absorption.
Catalog No. 50-274-1233