Learn More
Medchemexpress LLC TrkA Canine HEK293 100ug
SDP
Catalog No. 5000180704
Encompass_Preferred
TrkA Protein (isoform 3) belongs to the nerve growth factor receptor family and can promote angiogenesis and has carcinogenic activity when overexpressed TrkA Protein (isoform 3) antagonizes the anti-proliferative NGF-NTRK1 signaling that promotes the differentiation of neuronal precursors TrkA Protein Canine (HEK293 His) is the recombinant canine-derived TrkA protein expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-80704 Supplier Medchemexpress LLC Supplier No. HYP73456100UG
TrkA Protein (isoform 3) belongs to the nerve growth factor receptor family and can promote angiogenesis and has carcinogenic activity when overexpressed TrkA Protein (isoform 3) antagonizes the anti-proliferative NGF-NTRK1 signaling that promotes the differentiation of neuronal precursors TrkA Protein Canine (HEK293 His) is the recombinant canine-derived TrkA protein expressed by HEK293 with N-His labeled tag