Learn More
Sino Biological Recombinant Human TrkA Protein (A608D, GST Tag), Active
Catalog No. 502574038
Encompass_Preferred
Recombinant human TRKA (A608D) (440-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4038 Supplier Sino Biological Supplier No. N1612DG500
Recombinant human TRKA (A608D) (440-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.