Learn More
ACROBiosystems ACROBiosystems Human TrkA / NTRK1 (192-402) Protein, Mouse IgG2a Fc Tag
Catalog No. 501957096
Encompass_Preferred
Recombinant Protein;100UG;Human TrkA (192-402), Mouse IgG2a Fc Tag (TRA-H5253) is expressed from human 293 cells (HEK293). It contains AA Gly 192 - Val 402 (Accession # P04629-2).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 49.1 kDa. The protein migrates as 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7096 Supplier ACROBiosystems Supplier No. TRAH5253100UG
Recombinant Protein;100UG;Human TrkA (192-402), Mouse IgG2a Fc Tag (TRA-H5253) is expressed from human 293 cells (HEK293). It contains AA Gly 192 - Val 402 (Accession # P04629-2).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 49.1 kDa. The protein migrates as 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.