Learn More
ACROBiosystems ACROBiosystems Human TrkA / NTRK1 (33-417) Protein, Mouse IgG2a Fc Tag
Catalog No. 501957089
Encompass_Preferred
Recombinant Protein;1MG;Human TrkA (33-417), Mouse IgG2a Fc Tag (TRA-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 33 - Gly 417 (Accession # P04629-2).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 68.2 kDa. The protein migrates as 80-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7089 Supplier ACROBiosystems Supplier No. TRAH52591MG
Recombinant Protein;1MG;Human TrkA (33-417), Mouse IgG2a Fc Tag (TRA-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 33 - Gly 417 (Accession # P04629-2).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 68.2 kDa. The protein migrates as 80-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.