Learn More
Medchemexpress LLC TrkA Rat HEK293 H 10ug
SDP
Catalog No. 5000180607
Encompass_Preferred
The TrkA protein is a receptor tyrosine kinase that is critical in central and peripheral nervous system development regulating neuronal proliferation differentiation and survival As a high-affinity NGF receptor TrkA undergoes homodimerization and autophosphorylation upon ligand binding activating downstream effectors (SHC1 FRS2 SH2B1 SH2B2 PLCG1) cascade TrkA Protein Rat (HEK293 His) is the recombinant rat-derived TrkA protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-80607 Supplier Medchemexpress LLC Supplier No. HYP7346010UG
The TrkA protein is a receptor tyrosine kinase that is critical in central and peripheral nervous system development regulating neuronal proliferation differentiation and survival As a high-affinity NGF receptor TrkA undergoes homodimerization and autophosphorylation upon ligand binding activating downstream effectors (SHC1 FRS2 SH2B1 SH2B2 PLCG1) cascade TrkA Protein Rat (HEK293 His) is the recombinant rat-derived TrkA protein expressed by HEK293 with C-His labeled tag