Learn More
Eurofins Discoverx TrkB, active, 10 µg. (14-507)
Supplier:  Eurofins Discoverx 14507
Encompass
N-terminal 6xHis-tagged recombinant, human TrkB amino acids 455-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8887