Learn More
Sino Biological Recombinant Human NEK10 Protein (GST Tag), Active
Supplier: Sino Biological N1211G500
Recombinant human NEK10 (266-895) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.