Learn More
Medchemexpress LLC TrkB Human HEK293 100ug
SDP
Catalog No. 5000180490
Encompass_Preferred
The TrkB protein is a key receptor tyrosine kinase that plays a key role in central and peripheral nervous system development It regulates neuronal survival proliferation migration differentiation synapse formation and plasticity TrkB Protein Human (HEK293 His-Fc) is the recombinant human-derived TrkB protein expressed by HEK293 with C-hFc C-10 His labeled tag
Catalog No. 50-001-80490 Supplier Medchemexpress LLC Supplier No. HYP73462100UG
The TrkB protein is a key receptor tyrosine kinase that plays a key role in central and peripheral nervous system development It regulates neuronal survival proliferation migration differentiation synapse formation and plasticity TrkB Protein Human (HEK293 His-Fc) is the recombinant human-derived TrkB protein expressed by HEK293 with C-hFc C-10 His labeled tag