Learn More
Novus Biologicals™ TrkB [p Tyr816] Antibody Blocking Peptide

Specifications
Specifications
| Gene ID (Entrez) | 4915 |
| Species | Rat |
| Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-03499 |
| Gene Alias | BDNF/NT-3 growth factors receptor, EC 2.7.10, EC 2.7.10.1, GP145-TrkB, Neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, trk-B, TrkB tyrosine kinase, TRKBBDNF-tropomyosine receptor kinase B, tyrosine kinase receptor B |
| Gene Symbol | NTRK2 |
| Quantity | 0.05 mg |
| Research Category | Phospho Specific, Protein Kinase, Signal Transduction |
| Storage Buffer | From PBS. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.