Learn More
ACROBiosystems Recombinant Protein;Human TrkB / NTRK2 Protein, Fc Tag;HEK293;1MG;NT2,NTRK2,TRKB,NTRK-2,TRK-B,GP145-TrkB
Catalog No. 501658073
Encompass_Preferred
Recombinant Protein;Human TrkB / NTRK2 Protein, Fc Tag;1MG;Human TrkB, Fc Tag (NT2-H5254) is expressed from human 293 cells (HEK293). It contains AA Cys 32 - His 430 (Accession # AAH31835).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 70.4 kDa. The protein migrates as 95-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8073 Supplier ACROBiosystems Supplier No. NT2H52541MG
Recombinant Protein;Human TrkB / NTRK2 Protein, Fc Tag;1MG;Human TrkB, Fc Tag (NT2-H5254) is expressed from human 293 cells (HEK293). It contains AA Cys 32 - His 430 (Accession # AAH31835).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 70.4 kDa. The protein migrates as 95-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.