Learn More
ACROBiosystems Recombinant Protein;Human TrkB / NTRK2 Protein;HEK293;100UG;NT2,NTRK2,TRKB,NTRK-2,TRK-B,GP145-TrkB
Catalog No. 501658065
Encompass_Preferred
Recombinant Protein;Human TrkB / NTRK2 Protein;100UG;Human TrkB, His Tag (NT2-H5228) is expressed from human 293 cells (HEK293). It contains AA Cys 32 - His 430 (Accession # AAH31835).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 45 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8065 Supplier ACROBiosystems Supplier No. NT2H5228100UG
Recombinant Protein;Human TrkB / NTRK2 Protein;100UG;Human TrkB, His Tag (NT2-H5228) is expressed from human 293 cells (HEK293). It contains AA Cys 32 - His 430 (Accession # AAH31835).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 45 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.