Learn More
Medchemexpress LLC TrkC Human HEK293 50ug
SDP
Catalog No. 5000180349
Encompass_Preferred
TrkC Protein a receptor tyrosine kinase crucially contributes to nervous system and potential heart development Upon NTF3/neurotrophin-3 binding TrkC undergoes autophosphorylation activating pathways like phosphatidylinositol 3-kinase/AKT and MAPK These pathways regulate cell survival and differentiation emphasizing TrkC s pivotal role in essential cellular processes during development TrkC Protein Human (HEK293 His) is the recombinant human-derived TrkC protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-80349 Supplier Medchemexpress LLC Supplier No. HYP7346550UG
TrkC Protein a receptor tyrosine kinase crucially contributes to nervous system and potential heart development Upon NTF3/neurotrophin-3 binding TrkC undergoes autophosphorylation activating pathways like phosphatidylinositol 3-kinase/AKT and MAPK These pathways regulate cell survival and differentiation emphasizing TrkC s pivotal role in essential cellular processes during development TrkC Protein Human (HEK293 His) is the recombinant human-derived TrkC protein expressed by HEK293 with C-His labeled tag