Learn More
Sino Biological Recombinant Human TPR & TRKA (TRK-T1) Protein (GST Tag), Active
Catalog No. 502574018
Encompass_Preferred
Recombinant human fusion protein TPR (1-191)-TRKA (399-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4018 Supplier Sino Biological Supplier No. N1619BG500
Recombinant human fusion protein TPR (1-191)-TRKA (399-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.