Learn More
Carnabio Usa Inc TRKC(NTRK3)/10UG/CATALYTIC
Supplier:  Carnabio Usa Inc 0819710
Encompass
TRKC(NTRK3), Catalytic domain, Wild type, Amino Acid 456-825, NP_002521.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-1012