Learn More
Enzo Life Sciences Polyinosinic-polycytidylic acid . potassium salt (TLRGRADE) (synthetic), (5mg), CAS Number: 31852-29-6
SDP
Catalog No. NC9871421
Encompass_Preferred
Activator for TLR3 and MDA5. Specific activator for TLR3 (Toll-like receptor 3) and MDA5/Helicard. Alternative Name: poly(I:C) . K. Formula: (C10H10N4NaO7P)x • (C9H11N3NaO7P)x. CAS Number: 31852-29-6. Use/Stability: Aqueous stock solution is stable for 1 day when stored at +4°C. Long Term Storage: +4°C
Catalog No. NC9871421 Supplier Enzo Life Sciences Supplier No. ALX746021M005
Activator for TLR3 and MDA5. Specific activator for TLR3 (Toll-like receptor 3) and MDA5/Helicard. Alternative Name: poly(I:C) . K. Formula: (C10H10N4NaO7P)x • (C9H11N3NaO7P)x. CAS Number: 31852-29-6. Use/Stability: Aqueous stock solution is stable for 1 day when stored at +4°C. Long Term Storage: +4°C