Learn More
Aviva Systems Biology Corp TRMT11 Antibody - N-terminal region (ARP48526_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48526_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRMT11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRMT11. Alternate names: C6orf75, MDS024, TRM11, dJ187J11, dJ187J11.2, TRMT11-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59849