Learn More
Aviva Systems Biology Corp TRMT6 Antibody - middle region (ARP56780_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56780_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRMT6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human TRMT6. Alternate names: CGI-09, GCD10, Gcd10p, MGC5029ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-211