Learn More
Cayman Chemical Trandolapril, Purity: ≥98%, CAS: 87679-37-6, 100 mg
SDP
Catalog No. 502955243
Encompass_Preferred
A prodrug form of trandolaprilat; inhibits angiotensin I-induced pressor responses in rats and dogs (ID50s = 13.1 and 21.1 μg/kg, respectively); potentiates the bradykinin-induced depressor response in rats (ED50 = 5.5 μg/kg); decreases systolic and diastolic arterial pressure in dogs at 0.1, 0.3, and 1 mg/kg; decreases MAP and LVEDP in a rat model of left coronary artery occlusion-induced heart failure at 3 mg/kg; increases survival in stroke-prone spontaneously hypertensive rats fed a high-salt diet
Catalog No. 50-295-5243 Supplier Cayman Chemical Supplier No. 13522100 MG
A prodrug form of trandolaprilat; inhibits angiotensin I-induced pressor responses in rats and dogs (ID50s = 13.1 and 21.1 μg/kg, respectively); potentiates the bradykinin-induced depressor response in rats (ED50 = 5.5 μg/kg); decreases systolic and diastolic arterial pressure in dogs at 0.1, 0.3, and 1 mg/kg; decreases MAP and LVEDP in a rat model of left coronary artery occlusion-induced heart failure at 3 mg/kg; increases survival in stroke-prone spontaneously hypertensive rats fed a high-salt diet