Learn More
Cayman Chemical trans-2-Decenoic Acid, Purity: ≥95%, CAS: 334-49-6, 10 g
SDP
Catalog No. 502966247
Encompass_Preferred
An unsaturated fatty acid found in royal jelly produced from the hypopharyngeal and mandibular gland secretions of honeybees; isolated in conjunction with 10-HDA (Item No. 10976) and both demonstrate weak estrogenic activity, inhibiting binding of 17β-estradiol (Item No. ISO60155) to estrogen receptor β
Catalog No. 50-296-6247 Supplier Cayman Chemical Supplier No. 1788810 G
An unsaturated fatty acid found in royal jelly produced from the hypopharyngeal and mandibular gland secretions of honeybees; isolated in conjunction with 10-HDA (Item No. 10976) and both demonstrate weak estrogenic activity, inhibiting binding of 17β-estradiol (Item No. ISO60155) to estrogen receptor β