Learn More
Cayman Chemical trns-2-DecenoIc AcId 5g
SDP
Catalog No. 502979803
Encompass_Preferred
An unsaturated fatty acid found in royal jelly produced from the hypopharyngeal and mandibular gland secretions of honeybees; isolated in conjunction with 10-HDA (Item No. 10976) and both demonstrate weak estrogenic activity, inhibiting binding of 17β-estradiol (Item No. ISO60155) to estrogen receptor β
Catalog No. 50-297-9803 Supplier Cayman Chemical Supplier No. 178885 G
An unsaturated fatty acid found in royal jelly produced from the hypopharyngeal and mandibular gland secretions of honeybees; isolated in conjunction with 10-HDA (Item No. 10976) and both demonstrate weak estrogenic activity, inhibiting binding of 17β-estradiol (Item No. ISO60155) to estrogen receptor β