Learn More
Cayman Chemical trans-3'-Hydroxycotinine, Purity: ≥95%, CAS: 34834-67-8, 10 mg
SDP
Catalog No. 502968839
Encompass_Preferred
A product of CYP2A6 metabolism of the primary nicotine metabolite, cotinine; as a biomarker of nicotine metabolism
Catalog No. 50-296-8839 Supplier Cayman Chemical Supplier No. 1610010 MG
A product of CYP2A6 metabolism of the primary nicotine metabolite, cotinine; as a biomarker of nicotine metabolism