Learn More
Cayman Chemical trans-carboxy Glimepiride, Purity: ≥95%, CAS: 1217739-04-2, 1 mg
SDP
Catalog No. 502950355
Encompass_Preferred
A metabolite of glimepiride; formed from glimepiride in a two-step process mediated by CYP2C9 and cytosolic enzymes
Catalog No. 50-295-0355 Supplier Cayman Chemical Supplier No. 342771 MG
A metabolite of glimepiride; formed from glimepiride in a two-step process mediated by CYP2C9 and cytosolic enzymes