Learn More
Cayman Chemical trns-ClovamIde 10mg
SDP
Catalog No. 502970727
Encompass_Preferred
A naturally occurring caffeoyl conjugate with antioxidant and antiradical properties; demonstrates neuroprotective effects (EC50s = 0.9-3.7 µM) in several in vitro models of neuronal death
Catalog No. 50-297-0727 Supplier Cayman Chemical Supplier No. 1613810 MG
A naturally occurring caffeoyl conjugate with antioxidant and antiradical properties; demonstrates neuroprotective effects (EC50s = 0.9-3.7 µM) in several in vitro models of neuronal death