Learn More
Cayman Chemical -trns-GK563 10mg
SDP
Catalog No. 502933128
Encompass_Preferred
A GVIA iPLA2 inhibitor (IC50 = 1 nM); selective for GVIA iPLA2 over GVIA cPLA2 (IC50 = 22 μM); inhibits apoptosis induced by IL-1β and IFN-γ in INS-1 β-cells at 1 and 10 μM
Catalog No. 50-293-3128 Supplier Cayman Chemical Supplier No. 2573210 MG
A GVIA iPLA2 inhibitor (IC50 = 1 nM); selective for GVIA iPLA2 over GVIA cPLA2 (IC50 = 22 μM); inhibits apoptosis induced by IL-1β and IFN-γ in INS-1 β-cells at 1 and 10 μM