Learn More
Cayman Chemical trns-Indole-3-acrylIc AcId 1g
SDP
Catalog No. 502943230
Encompass_Preferred
A metabolite of L-tryptophan; increases the levels of mRNA encoding mucin 2 and Il-10, as well as decreases mRNA encoding Tnf-α, in primary mouse large intestinal spheroids co-cultured with LPS-stimulated primary mouse BMDMs; has been used as a precursor in the synthesis of anticancer agents and tubulin polymerization inhibitors, as well as used as a reactive chromophore probe to assess the α and β subunit conformational changes of S. typhimurium tryptophan synthase in tryptophan biosynthesis
Catalog No. 50-294-3230 Supplier Cayman Chemical Supplier No. 406551 G
A metabolite of L-tryptophan; increases the levels of mRNA encoding mucin 2 and Il-10, as well as decreases mRNA encoding Tnf-α, in primary mouse large intestinal spheroids co-cultured with LPS-stimulated primary mouse BMDMs; has been used as a precursor in the synthesis of anticancer agents and tubulin polymerization inhibitors, as well as used as a reactive chromophore probe to assess the α and β subunit conformational changes of S. typhimurium tryptophan synthase in tryptophan biosynthesis