Learn More
Cayman Chemical trans-Ned-19, Purity: ≥98%, CAS: 1354235-96-3, 10 mg
SDP
Catalog No. 502967637
Encompass_Preferred
A structural analog of NAADP that blocks NAADP-dependent calcium release (IC50 = 6 nM) as well as NAADP binding to sea urchin egg homogenate (IC50 = 0.4 nM); does not affect either IP3-mediated calcium release or cyclic ADP-ribose-mediated calcium release
Catalog No. 50-296-7637 Supplier Cayman Chemical Supplier No. 1752710 MG
A structural analog of NAADP that blocks NAADP-dependent calcium release (IC50 = 6 nM) as well as NAADP binding to sea urchin egg homogenate (IC50 = 0.4 nM); does not affect either IP3-mediated calcium release or cyclic ADP-ribose-mediated calcium release