Learn More
Cayman Chemical trans-Nerolidol, Purity: ≥98%, CAS: 40716-66-3, 100 mg
SDP
Catalog No. 502936610
Encompass_Preferred
A sesquiterpene with diverse biological activities; inhibits the growth of S. aureus, B. subtilis, E. coli, and S. cerevisiae; reduces viability of CaCo-2 adenocarcinoma cells (IC50 = 28.7 mg/L); insecticidal against A. aegypti (LC50 = 9 mg/L)
Catalog No. 50-293-6610 Supplier Cayman Chemical Supplier No. 25778100 MG
A sesquiterpene with diverse biological activities; inhibits the growth of S. aureus, B. subtilis, E. coli, and S. cerevisiae; reduces viability of CaCo-2 adenocarcinoma cells (IC50 = 28.7 mg/L); insecticidal against A. aegypti (LC50 = 9 mg/L)