Learn More
Cayman Chemical trns-PetroselInIc AcId 100mg
SDP
Catalog No. 502983104
Encompass_Preferred
A monounsaturated fatty acid; the isomer of oleic acid and trans isomer of petroselinic acid; has been found in cow, goat, and ewe milk fat as well as human breast milk; increases cellular content of triacylglycerols and cholesterol esters and upregulates transcription of genes involved in fatty acid synthesis, including SREBP-1c, ACACA, FASN, and SCD1, and cholesterol synthesis, including HMGCR, HMGCS1, FDFT1, and SREBP-2, in HepG2 cells at 100 μM
Catalog No. 50-298-3104 Supplier Cayman Chemical Supplier No. 20026100 MG
A monounsaturated fatty acid; the isomer of oleic acid and trans isomer of petroselinic acid; has been found in cow, goat, and ewe milk fat as well as human breast milk; increases cellular content of triacylglycerols and cholesterol esters and upregulates transcription of genes involved in fatty acid synthesis, including SREBP-1c, ACACA, FASN, and SCD1, and cholesterol synthesis, including HMGCR, HMGCS1, FDFT1, and SREBP-2, in HepG2 cells at 100 μM