Learn More
Cayman Chemical trns-Resveratrol-d4 100ug
SDP
Catalog No. 502955883
Encompass_Preferred
An internal standard for the quantification of trans-resveratrol by GC- or LC-MS
Catalog No. 50-295-5883 Supplier Cayman Chemical Supplier No. 13130100 UG
Add to Cart
Add to Cart
An internal standard for the quantification of trans-resveratrol by GC- or LC-MS