Learn More
Cayman Chemical trans-Resveratrol-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1089051-56-8, 1 mg
SDP
Catalog No. 502953094
Encompass_Preferred
An internal standard for the quantification of trans-resveratrol by GC- or LC-MS
Catalog No. 50-295-3094 Supplier Cayman Chemical Supplier No. 131301 MG
An internal standard for the quantification of trans-resveratrol by GC- or LC-MS