Learn More
Cayman Chemical trans-Vaccenic Acid, Purity: ≥98%, CAS: 693-72-1, 100 mg
SDP
Catalog No. 502972831
Encompass_Preferred
An ω-7 fatty acid; dietary administration reduces total body fat, mesenteric fat, and adipocyte size, increases inguinal fat mass, and decreases intestinal and hepatic triglyceride secretion in a rat model of obesity with features of metabolic syndrome at 1% w/w; decreases hepatocellular ballooning and steatosis in the same model; dietary administration of a butter enriched with trans-vaccenic acid decreases serum cholesterol levels and the formation of aortic atherosclerotic lesions in Ldlr-/- mice
Catalog No. 50-297-2831 Supplier Cayman Chemical Supplier No. 15301100 MG
An ω-7 fatty acid; dietary administration reduces total body fat, mesenteric fat, and adipocyte size, increases inguinal fat mass, and decreases intestinal and hepatic triglyceride secretion in a rat model of obesity with features of metabolic syndrome at 1% w/w; decreases hepatocellular ballooning and steatosis in the same model; dietary administration of a butter enriched with trans-vaccenic acid decreases serum cholesterol levels and the formation of aortic atherosclerotic lesions in Ldlr-/- mice