Learn More
Cayman Chemical trns-VaccenIc AcId-d13 10mg
SDP
Catalog No. 502940082
Encompass_Preferred
An internal standard for the quantification of trans-vaccenic acid by GC- or LC-MS
Catalog No. 50-294-0082 Supplier Cayman Chemical Supplier No. 2771710 MG
An internal standard for the quantification of trans-vaccenic acid by GC- or LC-MS