Learn More
Cayman Chemical trns-VaccenIc AcId-d13 1mg
SDP
Catalog No. 502939486
Encompass_Preferred
An internal standard for the quantification of trans-vaccenic acid by GC- or LC-MS
Catalog No. 50-293-9486 Supplier Cayman Chemical Supplier No. 277171 MG
An internal standard for the quantification of trans-vaccenic acid by GC- or LC-MS