Learn More
Cayman Chemical trns-VaccenIc AcId-d13 25mg
SDP
Catalog No. 502937945
Encompass_Preferred
An internal standard for the quantification of trans-vaccenic acid by GC- or LC-MS
Catalog No. 50-293-7945 Supplier Cayman Chemical Supplier No. 2771725 MG
An internal standard for the quantification of trans-vaccenic acid by GC- or LC-MS