Learn More
Cayman Chemical trns-ZeatIn 100mg
SDP
Catalog No. 502951885
Encompass_Preferred
A cytokinin plant growth regulator; binds to AHK3 (KD = 1.3 nM); increases chlorophyll levels in etiolated Cucumus sativus cotyledons in a concentration-dependent manner; increases callus growth and shoot formation in N. tabacum calluses at 5 and 50 µM; reduces production of ROS induced by Aβ25-35 in PC12 cells at 25-100 µM; scopolamine-induced spontaneous alternations in the Y-maze, indicating enhanced spatial memory, in mice at 1.5, 3, and 4.5 mg/kg per day
Catalog No. 50-295-1885 Supplier Cayman Chemical Supplier No. 13226100 MG
A cytokinin plant growth regulator; binds to AHK3 (KD = 1.3 nM); increases chlorophyll levels in etiolated Cucumus sativus cotyledons in a concentration-dependent manner; increases callus growth and shoot formation in N. tabacum calluses at 5 and 50 µM; reduces production of ROS induced by Aβ25-35 in PC12 cells at 25-100 µM; scopolamine-induced spontaneous alternations in the Y-maze, indicating enhanced spatial memory, in mice at 1.5, 3, and 4.5 mg/kg per day