Learn More
Cayman Chemical trns-ZeatIn RIbosIde 100mg
SDP
Catalog No. 502988622
Encompass_Preferred
A cytokinin; suppresses hypocotyl adventitious root formation in cucumber plants from 0.001-1 µM; increases the activity of SOD, APX, CAT, and POD in the leaves, as well as improves turf quality and delays leaf wilting in a creeping bentgrass model of drought stress at 10 µM
Catalog No. 50-298-8622 Supplier Cayman Chemical Supplier No. 30747100 MG
A cytokinin; suppresses hypocotyl adventitious root formation in cucumber plants from 0.001-1 µM; increases the activity of SOD, APX, CAT, and POD in the leaves, as well as improves turf quality and delays leaf wilting in a creeping bentgrass model of drought stress at 10 µM