Learn More
Cayman Chemical trans-Zeatin Riboside, Purity: ≥98%, CAS: 6025-53-2, 50 mg
SDP
Catalog No. 502988805
Encompass_Preferred
A cytokinin; suppresses hypocotyl adventitious root formation in cucumber plants from 0.001-1 µM; increases the activity of SOD, APX, CAT, and POD in the leaves, as well as improves turf quality and delays leaf wilting in a creeping bentgrass model of drought stress at 10 µM
Catalog No. 50-298-8805 Supplier Cayman Chemical Supplier No. 3074750 MG
A cytokinin; suppresses hypocotyl adventitious root formation in cucumber plants from 0.001-1 µM; increases the activity of SOD, APX, CAT, and POD in the leaves, as well as improves turf quality and delays leaf wilting in a creeping bentgrass model of drought stress at 10 µM