Learn More
Cayman Chemical (±)-trans-1,2-Bis(2-mercaptoacetamido)cyclohexane, Purity: ≥98%, CAS: 257641-01-3, 100 mg
SDP
Catalog No. 502966249
Encompass_Preferred
A dithiol used, both in vitro and in vivo, to promote the correct folding of proteins that require disulfide bonds for functionality
Catalog No. 50-296-6249 Supplier Cayman Chemical Supplier No. 17249100 MG
A dithiol used, both in vitro and in vivo, to promote the correct folding of proteins that require disulfide bonds for functionality