Learn More
Cayman Chemical trns1 2BIs 2mercaptoa 10mg
SDP
Catalog No. 502966844
Encompass_Preferred
A dithiol used, both in vitro and in vivo, to promote the correct folding of proteins that require disulfide bonds for functionality
Catalog No. 50-296-6844 Supplier Cayman Chemical Supplier No. 1724910 MG
A dithiol used, both in vitro and in vivo, to promote the correct folding of proteins that require disulfide bonds for functionality