Learn More
Cayman Chemical (±)-trans-1,2-Bis(2-mercaptoacetamido)cyclohexane, Purity: ≥98%, CAS: 257641-01-3, 50 mg
SDP
Catalog No. 502967492
Encompass_Preferred
A dithiol used, both in vitro and in vivo, to promote the correct folding of proteins that require disulfide bonds for functionality
Catalog No. 50-296-7492 Supplier Cayman Chemical Supplier No. 1724950 MG
A dithiol used, both in vitro and in vivo, to promote the correct folding of proteins that require disulfide bonds for functionality