Learn More
Cayman Chemical trns10HeptadecenoIc AcId 10mg
SDP
Catalog No. 502986056
Encompass_Preferred
A monounsaturated fatty acid; a minor constituent of ruminant fats; found in the monounsaturated fatty acid pool of C. atrox
Catalog No. 50-298-6056 Supplier Cayman Chemical Supplier No. 2246810 MG
A monounsaturated fatty acid; a minor constituent of ruminant fats; found in the monounsaturated fatty acid pool of C. atrox