Learn More
Cayman Chemical trans-10-Heptadecenoic Acid, Purity: ≥98%, CAS: 126761-43-1, 50 mg
SDP
Catalog No. 502986949
Encompass_Preferred
A monounsaturated fatty acid; a minor constituent of ruminant fats; found in the monounsaturated fatty acid pool of C. atrox
Catalog No. 50-298-6949 Supplier Cayman Chemical Supplier No. 2246850 MG
A monounsaturated fatty acid; a minor constituent of ruminant fats; found in the monounsaturated fatty acid pool of C. atrox